Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290437.1 | NP_001277366.1 | 10 - 11 | 2153 | 69 | |
GenBank mRNA | AK085439.1 | - | 10 - 11 | 2158 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons |