Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK036623.1 | - | 5 - 6 | 505 | 86 | |
AK151178.1 | - | 5 - 6 | 489 | 86 | ||
AK153039.1 | - | 5 - 6 | 489 | 86 | ||
AK169948.1 | - | 5 - 6 | 507 | 86 | ||
AK171063.1 | - | 5 - 6 | 491 | 86 | ||
AY753194.1 | - | 3 - 4 | 257 | 86 | ||
BC028998.1 | - | 5 - 6 | 497 | 5356085 | 86 | |
DQ295832.1 | - | 3 - 4 | 257 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |