Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172262.3 | NP_758466.1 | 4 - 5 | 494 | 77 | |
GenBank mRNA | BC023917.1 | - | 4 - 5 | 435 | 5323227 | 77 |
CF531857.1 | - | 4 - 5 | 442 | 77 | ||
CF733387.1 | - | 3 - 4 | 257 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |