Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081132.1 | NP_001074601.1 | 16 - 17 | 3267 | 125 | |
GenBank mRNA | BC138358.1 | - | 16 - 17 | 3230 | 8861378 | 125 |
BC138359.1 | - | 16 - 17 | 3230 | 8861379 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |