Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028804.1 | NP_083080.1 | 1 - 2 | 528 | 70 | |
GenBank mRNA | AK009833.1 | - | 1 - 2 | 372 | 70 | |
BC058616.1 | - | 1 - 2 | 528 | 5686139 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |