Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081349.2 | NP_001074818.2 | 10 - 11 | 1332 | 78 | |
NM_001083809.1 | NP_001077278.1 | 9 - 10 | 1271 | 78 | ||
NM_024497.1 | NP_078773.1 | 10 - 11 | 1230 | 78 | ||
GenBank mRNA | AB103034.1 | - | 10 - 11 | 1301 | 78 | |
AK011417.1 | - | 10 - 11 | 1330 | 78 | ||
AK031215.1 | - | 9 - 10 | 1274 | 78 | ||
AK046758.1 | - | 10 - 11 | 1324 | 78 | ||
AK165374.1 | - | 10 - 11 | 1324 | 78 | ||
BC053747.1 | - | 10 - 11 | 1129 | 6330614 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |