Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146251.4 | NP_666363.3 | 14 - 15 | 1713 | 72 | |
GenBank mRNA | AK038505.1 | - | 14 - 15 | 1599 | 72 | |
AK088362.1 | - | 14 - 15 | 1659 | 72 | ||
AK148380.1 | - | 14 - 15 | 1687 | 72 | ||
AK170417.1 | - | 14 - 15 | 1659 | 72 | ||
BC021312.1 | - | 13 - 14 | 1747 | 72 | ||
BC025621.1 | - | 10 - 11 | 1126 | 72 | ||
BC141365.1 | - | 14 - 15 | 1497 | 9055978 | 72 | |
BC141366.1 | - | 14 - 15 | 1497 | 9055979 | 72 | |
EU293208.1 | - | 13 - 14 | 1384 | 72 | ||
EU368675.1 | - | 13 - 14 | 1384 | 72 | ||
EU402948.2 | - | 13 - 14 | 1384 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |