Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009849.2 | NP_033979.2 | 4 - 5 | 646 | 66 | |
GenBank mRNA | AF042811.1 | - | 4 - 5 | 570 | 66 | |
AK002553.1 | - | 4 - 5 | 618 | 66 | ||
AK172281.1 | - | 4 - 5 | 594 | 66 | ||
AY376711.1 | - | 4 - 5 | 579 | 66 | ||
BC011241.1 | - | 4 - 5 | 578 | 3582550 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |