Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016716.4 | NP_057925.1 | 6 - 7 | 1008 | 145 | |
GenBank mRNA | AF129738.1 | - | 6 - 7 | 898 | 145 | |
AK129174.1 | - | 7 - 8 | 1396 | 145 | ||
BC027304.1 | - | 6 - 7 | 1033 | 3981170 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |