Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025664.5 | NP_079940.2 | 2 - 3 | 254 | 75 | |
GenBank mRNA | AK139802.1 | - | 2 - 3 | 288 | 75 | |
AK149739.1 | - | 2 - 3 | 268 | 75 | ||
AK149849.1 | - | 2 - 3 | 308 | 75 | ||
AK155830.1 | - | 289 | 75 | |||
AK157517.1 | - | 2 - 3 | 270 | 75 | ||
AK170339.1 | - | 270 | 75 | |||
AK202748.1 | - | 2 - 3 | 103 | 75 | ||
AK202820.1 | - | 2 - 3 | 103 | 75 | ||
AK208099.1 | - | 1 - 2 | 94 | 75 | ||
AK218282.1 | - | 2 - 3 | 103 | 75 | ||
BC014814.1 | - | 2 - 3 | 254 | 3257422 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |