Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019450.3 | NP_062323.1 | 1 - 2 | 121 | 83 | |
GenBank mRNA | AK004061.1 | - | 1 - 2 | 121 | 83 | |
AK224928.1 | - | 1 - 2 | 67 | 83 | ||
AK224929.1 | - | 1 - 2 | 67 | 83 | ||
AK224931.1 | - | 1 - 2 | 55 | 83 | ||
AK224932.1 | - | 1 - 2 | 68 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Probe spans exons |