Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027748.3 | NP_082024.2 | 1 - 2 | 379 | 66 | |
GenBank mRNA | AJ292189.1 | - | 1 - 2 | 170 | 66 | |
AK154372.1 | - | 1 - 2 | 388 | 66 | ||
BB621106.1 | - | 1 - 2 | 380 | 66 | ||
BC039989.1 | - | 1 - 2 | 335 | 66 | ||
BC089030.1 | - | 1 - 2 | 218 | 66 | ||
BC094546.1 | - | 1 - 2 | 336 | 66 | ||
BC137617.1 | - | 1 - 2 | 320 | 8860636 | 66 | |
BC137618.1 | - | 1 - 2 | 320 | 8860637 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |