Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081173.1 | NP_001074642.1 | 12 - 13 | 1589 | 79 | |
GenBank mRNA | AK122516.1 | - | 1 - 2 | 1404 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |