Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdkrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009747.2 | NP_033877.3 | 1 - 2 | 76 | 72 | |
GenBank mRNA | X78438.1 | - | 1 - 2 | 76 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mus musculus Inventoried Probe spans exons |