Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00018522.mm.1 |
NM_001105245.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018522.mm.1 |
NM_001105245.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018522.mm.1 |
NM_001105246.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018522.mm.1 |
NM_001105246.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018522.mm.1 |
TC0X00002982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018523.mm.1 |
NM_001105245.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018523.mm.1 |
NM_001105245.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018523.mm.1 |
NM_001105246.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018523.mm.1 |
NM_001105246.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018523.mm.1 |
TC0X00002982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018524.mm.1 |
NM_001105245.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018524.mm.1 |
NM_001105245.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018524.mm.1 |
NM_001105246.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018524.mm.1 |
NM_001105246.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018524.mm.1 |
TC0X00002982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018521.mm.1 |
NM_001105245.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018521.mm.1 |
NM_001105245.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018521.mm.1 |
NM_001105246.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018521.mm.1 |
NM_001105246.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018521.mm.1 |
TC0X00002982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018525.mm.1 |
NM_001105245.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018525.mm.1 |
NM_001105245.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018525.mm.1 |
NM_001105246.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018525.mm.1 |
NM_001105246.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018525.mm.1 |
TC0X00002982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018519.mm.1 |
NM_001105245.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018519.mm.1 |
NM_001105245.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018519.mm.1 |
NM_001105246.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018519.mm.1 |
NM_001105246.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018519.mm.1 |
TC0X00002982.mm.1 |
MTA TC |