Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800017197.mm.1 |
NM_001167981.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017197.mm.1 |
NM_001167981.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017197.mm.1 |
NM_144731.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017197.mm.1 |
NM_144731.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017197.mm.1 |
TC0800002256.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017199.mm.1 |
NM_001167981.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017199.mm.1 |
NM_001167981.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017199.mm.1 |
NM_144731.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017199.mm.1 |
NM_144731.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017199.mm.1 |
TC0800002256.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017198.mm.1 |
NM_001167981.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017198.mm.1 |
NM_001167981.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017198.mm.1 |
NM_144731.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017198.mm.1 |
NM_144731.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017198.mm.1 |
TC0800002256.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017196.mm.1 |
NM_001167981.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017196.mm.1 |
NM_001167981.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017196.mm.1 |
NM_144731.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017196.mm.1 |
NM_144731.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017196.mm.1 |
TC0800002256.mm.1 |
MTA TC |