Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3rf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021506.2 | NP_067481.2 | 1 - 2 | 175 | 81 | |
GenBank mRNA | AF030131.1 | - | 1 - 2 | 74 | 81 | |
AK088003.1 | - | 1 - 2 | 115 | 81 | ||
AK148137.1 | - | 1 - 2 | 103 | 81 | ||
AK173185.1 | - | 1 - 2 | 538 | 81 | ||
BC060113.1 | - | 1 - 2 | 137 | 6405370 | 81 | |
BC060696.1 | - | 1 - 2 | 137 | 6405370 | 81 | |
BY741307.1 | - | 1 - 2 | 101 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |