Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021322.2 | NP_067297.2 | 5 - 6 | 796 | 72 | |
GenBank mRNA | AJ271892.1 | - | 5 - 6 | 566 | 72 | |
AJ271893.1 | - | 5 - 6 | 566 | 72 | ||
AK035125.1 | - | 4 - 5 | 420 | 72 | ||
AK144821.1 | - | 5 - 6 | 601 | 72 | ||
AK152271.1 | - | 5 - 6 | 588 | 72 | ||
AK153161.1 | - | 5 - 6 | 588 | 72 | ||
AK164160.1 | - | 5 - 6 | 509 | 72 | ||
BC039272.1 | - | 5 - 6 | 596 | 5027600 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |