Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008823.3 | NP_032849.2 | 2 - 3 | 284 | 56 | |
GenBank mRNA | AK150212.1 | - | 2 - 3 | 245 | 56 | |
AK152782.1 | - | 2 - 3 | 235 | 56 | ||
AK154306.1 | - | 2 - 3 | 268 | 56 | ||
AK159291.1 | - | 2 - 3 | 285 | 56 | ||
AK159609.1 | - | 2 - 3 | 235 | 56 | ||
AK159853.1 | - | 2 - 3 | 235 | 56 | ||
AK171374.1 | - | 2 - 3 | 268 | 56 | ||
BC132369.1 | - | 2 - 3 | 268 | 40130646 | 56 | |
BC138306.1 | - | 2 - 3 | 268 | 8861326 | 56 | |
X12905.1 | - | 1 - 2 | 133 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |