Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025074.2 | NP_001020245.1 | 7 - 8 | 1380 | 67 | |
NM_001282961.1 | NP_001269890.1 | 7 - 8 | 1347 | 67 | ||
NM_008745.3 | NP_032771.1 | 7 - 8 | 1347 | 67 | ||
GenBank mRNA | AB377224.1 | - | 6 - 7 | 757 | 67 | |
AB377225.1 | - | 6 - 7 | 757 | 67 | ||
AK018789.1 | - | 7 - 8 | 1278 | 67 | ||
AK028827.1 | - | 7 - 8 | 1200 | 67 | ||
AK147391.1 | - | 7 - 8 | 1178 | 67 | ||
AK147441.1 | - | 6 - 7 | 1168 | 67 | ||
AK160789.1 | - | 7 - 8 | 1348 | 67 | ||
BC052014.1 | - | 6 - 7 | 1142 | 5707891 | 67 | |
KC855567.1 | - | 1 - 2 | 86 | 67 | ||
M33385.1 | - | 6 - 7 | 719 | 67 | ||
X17647.1 | - | 1230 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |