Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Avl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030235.1 | NP_084511.1 | 2 - 3 | 431 | 67 | |
GenBank mRNA | AK020003.1 | - | 2 - 3 | 408 | 67 | |
AK083605.1 | - | 2 - 3 | 387 | 67 | ||
AK122228.1 | - | 2 - 3 | 409 | 67 | ||
AK147365.1 | - | 2 - 3 | 433 | 67 | ||
BC051177.1 | - | 2 - 3 | 413 | 67 | ||
BC117878.1 | - | 2 - 3 | 393 | 40095961 | 67 | |
BC117879.1 | - | 2 - 3 | 393 | 67 | ||
CJ109208.1 | - | 2 - 3 | 433 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |