Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shisa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001174086.1 | NP_001167557.1 | 3 - 4 | 1321 | 85 | |
GenBank mRNA | BC030884.1 | - | 3 - 4 | 610 | 85 | |
GU479982.1 | - | 3 - 4 | 850 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |