Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrtc1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142690.1 | NP_001136162.1 | 5 - 6 | 773 | 68 |
Gene Symbol: Dmrtc1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142691.1 | NP_001136163.1 | 5 - 6 | 773 | 68 | |
GenBank mRNA | AK016891.1 | - | 5 - 6 | 774 | 68 | |
BC116811.1 | - | 4 - 5 | 407 | 40126130 | 68 | |
BC116813.1 | - | 4 - 5 | 407 | 40126132 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |