Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK142152.1 | - | 10 - 11 | 1198 | 109 | |
AK154645.1 | - | 7 - 8 | 3438 | 109 | ||
BC031484.1 | - | 10 - 11 | 1196 | 2654265 | 109 | |
BC044769.1 | - | 3 - 4 | 1141 | 109 | ||
BC046438.1 | - | 2 - 3 | 172 | 109 | ||
BC047372.1 | - | 10 - 11 | 1179 | 109 | ||
BC057597.1 | - | 8 - 9 | 2764 | 109 | ||
BC089379.1 | - | 10 - 11 | 1153 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |