Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145628.4 | NP_663603.3 | 1 - 2 | 210 | 94 | |
GenBank mRNA | AK031767.1 | - | 1 - 2 | 188 | 94 | |
AK166523.1 | - | 1 - 2 | 186 | 94 | ||
BY272535.1 | - | 1 - 2 | 211 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mus musculus Inventoried Probe spans exons |