Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009896.2 | NP_034026.1 | 1 - 2 | 103 | 133 | |
GenBank mRNA | AF120490.1 | - | 1 - 2 | 76 | 133 | |
AF180302.1 | - | 1 - 2 | 81 | 133 | ||
AK028632.1 | - | 104 | 133 | |||
BC132366.1 | - | 1 - 2 | 100 | 40130643 | 133 | |
BC132368.1 | - | 1 - 2 | 100 | 40130645 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |