Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032465.2 | NP_115854.2 | 9 - 10 | 1396 | 78 | |
GenBank mRNA | AK040539.1 | - | 9 - 10 | 1400 | 78 | |
AK154076.1 | - | 9 - 10 | 1366 | 78 | ||
AK156470.1 | - | 9 - 10 | 1397 | 78 | ||
AY029156.1 | - | 9 - 10 | 1254 | 78 | ||
BC052865.1 | - | 9 - 10 | 1293 | 30072864 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |