Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178720.4 | NP_848835.1 | 4 - 5 | 491 | 64 | |
GenBank mRNA | AK034628.1 | - | 4 - 5 | 492 | 64 | |
AK047981.1 | - | 3 - 4 | 401 | 64 | ||
AK052599.1 | - | 5 - 6 | 601 | 64 | ||
BC116254.1 | - | 4 - 5 | 460 | 40093631 | 64 | |
BC116255.1 | - | 4 - 5 | 460 | 40093632 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |