Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178720.4 | NP_848835.1 | 8 - 9 | 924 | 98 | |
GenBank mRNA | AK034628.1 | - | 8 - 9 | 925 | 98 | |
AK047981.1 | - | 7 - 8 | 834 | 98 | ||
AK052599.1 | - | 9 - 10 | 1034 | 98 | ||
BC116254.1 | - | 8 - 9 | 893 | 40093631 | 98 | |
BC116255.1 | - | 8 - 9 | 893 | 40093632 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |