Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mast3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199308.2 | NP_955012.2 | 17 - 18 | 2149 | 88 | |
GenBank mRNA | AK140821.1 | - | 17 - 18 | 2158 | 88 | |
AK155568.1 | - | 18 - 19 | 2121 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |