Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173440.2 | NP_775616.1 | 2 - 3 | 259 | 58 | |
GenBank mRNA | AK030495.1 | - | 2 - 3 | 244 | 58 | |
AK036273.1 | - | 2 - 3 | 260 | 58 | ||
AK041348.1 | - | 2 - 3 | 251 | 58 | ||
AK156233.1 | - | 2 - 3 | 238 | 58 | ||
AK157467.1 | - | 2 - 3 | 238 | 58 | ||
BC060232.1 | - | 1 - 2 | 57 | 6852643 | 58 | |
BC075638.1 | - | 2 - 3 | 136 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |