Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008941.3 | NP_032967.1 | 17 - 18 | 1969 | 137 | |
NM_178855.4 | NP_849186.2 | 16 - 17 | 1924 | 137 | ||
GenBank mRNA | AK033582.1 | - | 16 - 17 | 1926 | 137 | |
AK038356.1 | - | 8 - 9 | 918 | 137 | ||
BC117917.1 | - | 17 - 18 | 1968 | 40097367 | 137 | |
BC117918.1 | - | 17 - 18 | 1968 | 40097370 | 137 | |
U73378.1 | - | 17 - 18 | 1964 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |