Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crybg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174848.3 | NP_777273.2 | 7 - 8 | 2234 | 75 | |
GenBank mRNA | BC043118.1 | - | 7 - 8 | 2234 | 6408859 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |