Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081190.1 | NP_001074659.1 | 4 - 5 | 523 | 96 | |
GenBank mRNA | BC151105.1 | - | 4 - 5 | 573 | 9088018 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |