Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fopnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025345.2 | NP_079621.1 | 3 - 4 | 312 | 79 | |
GenBank mRNA | AK012206.1 | - | 3 - 4 | 307 | 79 | |
AK146884.1 | - | 3 - 4 | 338 | 79 | ||
AK153894.1 | - | 3 - 4 | 316 | 79 | ||
BC011300.1 | - | 3 - 4 | 312 | 3671642 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |