Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081255.1 | NP_001074724.1 | 7 - 8 | 1023 | 117 | |
GenBank mRNA | AK076529.1 | - | 7 - 8 | 1013 | 117 | |
AK155550.1 | - | 7 - 8 | 1021 | 117 | ||
BC151058.1 | - | 7 - 8 | 1001 | 9087971 | 117 | |
BC157924.1 | - | 7 - 8 | 1001 | 9088097 | 117 | |
BC172144.1 | - | 7 - 8 | 1001 | 9054814 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |