Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008782.2 | NP_032808.1 | 7 - 8 | 948 | 59 | |
GenBank mRNA | M97013.1 | - | 7 - 8 | 948 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |