Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033201.3 | NP_001028373.2 | 19 - 20 | 2243 | 68 | |
GenBank mRNA | AK133991.1 | - | 19 - 20 | 2199 | 68 | |
AK134334.1 | - | 19 - 20 | 2234 | 68 | ||
BC033473.1 | - | 18 - 19 | 1859 | 68 | ||
BC034658.1 | - | 16 - 17 | 1685 | 68 | ||
BC055777.1 | - | 16 - 17 | 1669 | 68 | ||
BC138898.1 | - | 19 - 20 | 2114 | 8861920 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |