Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028902.1 | NP_083178.1 | 2 - 3 | 220 | 132 | |
GenBank mRNA | AK016553.1 | - | 2 - 3 | 205 | 132 | |
DV650538.1 | - | 2 - 3 | 220 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |