Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1956737 |
NM_028902.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1956737 |
AK016553.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
153592 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
416405 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
202853 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
907431 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
568138 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
266573 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
793490 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
817291 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
335514 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
556244 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
163080 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
172137 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
557400 |
AK011733.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
557400 |
AK017354.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
557400 |
BC002278.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
557400 |
NM_024178.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
851593 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
497381 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
609677 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
511907 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
555636 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
259954 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
946482 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
766150 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
30095 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
652567 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572415 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
149780 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
646260 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
936048 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
287023 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
142613 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
12383 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
306796 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
363072 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
635531 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
145358 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
94207 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
506122 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
687167 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
707803 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
795878 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
488370 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
282856 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
331053 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
347669 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
5620 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
645973 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
619665 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
885600 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562316 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
542524 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
785968 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
299280 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
280536 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
169822 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
304460 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
730867 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
697471 |
AF011389.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
697471 |
AF190672.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
697471 |
AK149287.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
697471 |
BC014826.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
697471 |
NM_008106.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
354443 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
151481 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
140492 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
163222 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
2326 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
168218 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
111720 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
174691 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
161357 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
128236 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
174725 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P120331 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P114675 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1956737 |
NM_028902.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1956737 |
AK016553.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P114675 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P120331 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08759970 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08759991 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08760017 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03307319 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08759891 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08759922 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08760009 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P264411 |
NM_028902.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P264411 |
AK016553.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P125577 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P118133 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P114675 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P08760017 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P120331 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13215432 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31143117 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31143220 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13195386 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31143211 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13275691 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31143215 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31143120 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31143123 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31143119 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31143213 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13215433 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13160916 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31143218 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13195392 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13215435 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31143127 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13215440 |
- |
- |