Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252374.1 | NP_001239303.1 | 6 - 7 | 406 | 93 | |
NM_026004.3 | NP_080280.3 | 5 - 6 | 563 | 93 | ||
GenBank mRNA | AK011525.1 | - | 5 - 6 | 514 | 93 | |
AK011894.1 | - | 5 - 6 | 461 | 93 | ||
AK198650.1 | - | 2 - 3 | 142 | 93 | ||
BC038029.1 | - | 6 - 7 | 399 | 4018956 | 93 | |
CB571852.1 | - | 6 - 7 | 411 | 93 | ||
CX734167.1 | - | 3 - 4 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |