Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nktr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010918.2 | NP_035048.3 | 3 - 4 | 277 | 126 | |
GenBank mRNA | AK035582.1 | - | 2 - 3 | 178 | 126 | |
AK088056.1 | - | 3 - 4 | 216 | 126 | ||
AK144214.1 | - | 3 - 4 | 188 | 126 | ||
AK163916.1 | - | 3 - 4 | 290 | 126 | ||
BC048414.1 | - | 2 - 3 | 211 | 126 | ||
L04289.2 | - | 3 - 4 | 282 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |