Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113514.1 | NP_001106986.1 | 9 - 10 | 944 | 81 | |
NM_133721.2 | NP_598482.2 | 23 - 24 | 2765 | 81 | ||
GenBank mRNA | AJ344342.1 | - | 23 - 24 | 2768 | 81 | |
AK083987.1 | - | 8 - 9 | 784 | 81 | ||
AK139690.1 | - | 9 - 10 | 932 | 81 | ||
AK153585.1 | - | 21 - 22 | 2227 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Inventoried Probe spans exons |