Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015759.2 | NP_056574.2 | 5 - 6 | 899 | 72 | |
GenBank mRNA | AF017369.1 | - | 5 - 6 | 934 | 72 | |
AK051395.1 | - | 6 - 7 | 1422 | 72 | ||
AK157835.1 | - | 899 | 72 | |||
AK165413.1 | - | 5 - 6 | 1309 | 72 | ||
BC132340.1 | - | 3 - 4 | 718 | 40130617 | 72 | |
BC137954.1 | - | 3 - 4 | 719 | 8860973 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |