Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114087.2 | NP_001107559.1 | 4 - 5 | 375 | 74 | |
NM_001114088.2 | NP_001107560.1 | 4 - 5 | 375 | 74 | ||
NR_104287.1 | 4 - 5 | 375 | 74 | |||
GenBank mRNA | AK033091.1 | - | 367 | 74 | ||
AK049902.1 | - | 4 - 5 | 367 | 74 | ||
AK077998.1 | - | 4 - 5 | 367 | 74 | ||
AK128987.1 | - | 4 - 5 | 363 | 74 | ||
AK142933.1 | - | 4 - 5 | 358 | 74 | ||
AK167476.1 | - | 4 - 5 | 367 | 74 | ||
BC052698.1 | - | 4 - 5 | 346 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |