Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012324.2 | NP_001012324.1 | 1 - 2 | 197 | 88 | |
GenBank mRNA | AK038812.1 | - | 1 - 2 | 59 | 88 | |
AK040995.1 | - | 1 - 2 | 221 | 88 | ||
AK134746.1 | - | 1 - 2 | 151 | 88 | ||
AK163501.1 | - | 1 - 2 | 139 | 88 | ||
BC056468.1 | - | 1 - 2 | 90 | 88 | ||
BC065151.1 | - | 1 - 2 | 67 | 5708030 | 88 | |
BC089559.1 | - | 1 - 2 | 197 | 30295705 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |