Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lipi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252513.1 | NP_001239442.1 | 4 - 5 | 699 | 150 | |
GenBank mRNA | AK086576.1 | - | 4 - 5 | 702 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |