Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400004279.mm.1 |
NM_001195284.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004279.mm.1 |
NM_001195284.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004279.mm.1 |
TC0400000585.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400004282.mm.1 |
NM_001195284.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004282.mm.1 |
NM_001195284.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004282.mm.1 |
TC0400000585.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400004280.mm.1 |
NM_001195284.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004280.mm.1 |
NM_001195284.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004280.mm.1 |
TC0400000585.mm.1 |
MTA TC |