Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025900.2 | NP_080176.2 | 9 - 10 | 1215 | 141 | |
GenBank mRNA | AK007546.1 | - | 6 - 7 | 764 | 141 | |
AK033451.1 | - | 9 - 10 | 1105 | 141 | ||
AK141790.1 | - | 3 - 4 | 567 | 141 | ||
BC055451.1 | - | 9 - 10 | 1161 | 4164972 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |