Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcwpw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015254.1 | - | 1085 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |